Would you like to log in again?
Presaturated Wipes, FG Cleanwipes
Supplier: FG Cleanwipes
6LS70309APN40 6LS7030 6LS7030917 6LS7030685
76238-278CS 913.29 USD InStock
76238-278 76267-542 76238-280 76240-954
Presaturated Wipes, FG Cleanwipes
Wipes Disposable Wipes and Towels Presaturated Wipes
These wipes controls contamination in critical environment that depends on wet cleaning.
  • Variety of substrates available nonwoven and knitted
  • Custom saturant capabilities
  • Options for different types of sealed edges
  • Choice of pouch or canister
Benefits like reduced VOCs, minimized bulk liquid storage, safer to ship, store and use, consistency in application of solvent.
Certifications: Purity and cleanliness to support ISO 4 and ISO 5 environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty