Would you like to log in again?
Medical and Pharmaceutical Tapes, UltraTape Industries
Supplier: UltraTape
7160WH100-P3D
76238-300PK 1837.21 USD InStock
76238-300
Medical and Pharmaceutical Tapes, UltraTape Industries
Tapes
UltraTape’s unique tapes are designed specifically for Medical and Pharmaceutical applications. All tapes can be irradiated to meet the strict requirements of these industries.
  • Irradiated lot traceability and certification
  • Gap sealing tape for use during dry-fog/aerosol disinfection
  • Cuff Sealing Tape is perforated to allow the user to apply and tear the tape using one hand
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty