Would you like to log in again?
Air Dusters, ITW Chemtronics
Supplier: ITW Chemtronics
ES1017
76238-634CS 662.97 USD InStock
76238-634
Air Dusters, ITW Chemtronics
Sprays
Duster is an economical duster that removes particulates from electronic devices. This duster / cleaner is nonflammable, residue-free and won't scratch delicate surfaces.
  • Nonflammable
  • Regular cleaning strength
  • Zero VOC
  • Filtered to 0.2 microns
  • 100% ultra-pure HFC-134a
  • Leaves no residue
  • Safe on plastic
Removing debris from keyboards, office equipment and removing surface contamination from A/V equipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty