Would you like to log in again?
Stainless Steel Tweezers, Lindstrom Precision Tools
Supplier: Williams
LIND-TL-123-SA LIND-TL-574-SA LIND-TL-AA-SA 2AX-SA LIND-TL-00B-SA LIND-TL-571-SA TL-7A-SA LIND-TL-2A-SA
76238-926EA 129.22 USD InStock
76238-926 76238-878 76239-996 76240-090 76239-796 76240-092 76240-094 76239-998
Stainless Steel Tweezers, Lindstrom Precision Tools
Forceps
Lindström tweezers offer perfect balance, tip alignment, and symmetry. The non-magnetic stainless steel provides excellent resistance to many acids because of the high concentration of molybdenum.
  • Suitable for continuous high temperature use
  • Anti-magnetic
  • Anti-acid
High precision reverse action: These are tweezers especially intended for applications where a high precise self-closing action is required (e.g. microscopy). They permit holding parts without finger pressure.
High precision: Primarily suitable for delicate standard applications and precision work.
Boley: Popular tweezers for general purpose applications.
General purpose: Serrated tips and handles for a secure grip.
Component handling: Specifically designed tips hold delicate electronic components firmly and safely. These tweezers can handle many types of round components and electronic devices such as resistors and diodes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty