Would you like to log in again?
Universal Mounts, Production Basics
Supplier: Production Basics
PB-8640 8643 8640
11005-428EA 47.42 USD InStock
11005-428 76239-108 11005-430
Universal Mounts, Production Basics
Wall Hooks
Use with existing magnifiers, task lamps and document holders.
  • Made of epoxy powder coated steel
  • Attaches to Any C-Leg Back Frame or Other Bench Upright
Add uprights to mount accessories on RTW Lab Bench, Easy Lift Dual-Leg, PedSys Lab Bench, Basic Organizer, Accessory Tower or Easy Lift 4-post Lab Bench.
Ordering information: Standard accessory color is Black. Other standard frame color (Light Neutral/Almond, Blue, White, or Gray) available by request. Accessory is ready to mount on a C-Leg Lab Bench, Basic Organizer or Accessory Tower; or otherwise must purchase Uprights, sold separately, if mounting on a RTW Lab Bench, Easy Lift Dual-Leg, PedSys Lab Bench or Easy Lift 4-post Lab Bench.
Delivery information: Includes 1.27 cm (1/2") bore for mounting posts, Universal Mount (1), Parts and Hardware Packet (1).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty