Would you like to log in again?
Polyflow®-G Polypropylene Nominal-Rated Depth Cartridges, Parker Hannifin
Supplier: Parker
PG-10010-050-14 PG-10710-002-2 PG-10010-100-14 PG-10010-100-12 CO-10020 PG-10320-005-1 PG-10010-010-14 PG-10320-010-1 PG-10H20-005-1 PG-10110-050-11 PG-10620-010-0 PG-10110-100-11 PG-10330-005-1 PG-10310-010-2 PG-10310-002-1 PG-10620-002-0
76239-138CS 89.63 USD InStock
76239-138 76239-136 76239-158 76239-148 76239-152 76239-164 76239-142 76239-150 76239-162 76239-140 76239-156 76239-146 76239-154 76239-144 76240-414 76239-160
Polyflow®-G Polypropylene Nominal-Rated Depth Cartridges, Parker Hannifin
Filters Filter Cartridges
These are for economical prefiltration. Polyflow®-G depth media has been developed for a wide variety of general process applications from fluid clarification to general prefiltration. Its high dirt-loading, random-fiber depth media provides consistent particle retention.
  • Thermally bonded from 100% virgin polypropylene to ensure clean filtrates and excellent chemical and thermal compatibility
  • Overall reduction of filtration costs when compared to spunbonded, stringwound, and nominally-rated pleated prefilter cartridges
  • Longer filtration life reduces downtime due to fewer change-outs
High flow rate and long service life reduce processing time. Broad chemical compatibility allows use in most applications. Thermally bonded construction minimizes extractables for cleaner filtrate.
Applied for liquid clarification, general water filtration, beverage/wine clarifi cation, RO/DI prefiltration.
Effective filtration area: 0.33 m² (3.6 ft²) per 250 mm (10") cartridge.
Cartridge extractables: NVR <35 mg per 250 mm (10") cartridge.
Maximum differential pressure/temperature of forward: 80 psid (5.5 bar) at 75 °F (24 °C) and reverse: 40 psid (2.8 bar) at 75 °F (24 °C), 15 psid (1.0 bar) at 140 °F (60 °C).
Certifications: Meets USP specifi cations for Class VI-121 °C plastics criteria.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty