Would you like to log in again?
Bio Barb® Adapters, Bioconnexx
Supplier: BioConnex
PP-100-TC0375 PP-100-TC0500 PP150TC-0500HB PP-MINI-TC0500 PP-MINI-TC0250 PP-100-TC1000
76239-174EA 43.82 USD InStock
76239-174 76240-130 76245-284 76239-176 76240-128 76246-022
Bio Barb® Adapters, Bioconnexx
Fittings and Connectors
True bore design allows for full flow and no restriction of product transfer. Eliminates turbulence caused by the adapter reducees possible damage to the medium being transferred.
  • Slight raise at the port on the TC face ensuring proper capture of gasket
  • Produced from a polypropylene resin
Certifications: Polypropylene complies with FDA and USP Class VI requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty