Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Steriking® See-Through Self-Sealable Pouches, Healthmark Industries
Supplier: HEALTHMARK INDUSTRIES CO. TE
Steriking®
SS5A SS7 SS4 SS6 SS1 SS2
76239-310BX 321.87 USD InStock
76239-310 76239-304 76239-258 76239-302 76239-308 76239-306
Steriking® See-Through Self-Sealable Pouches, Healthmark Industries
Bags Sterilization Pouches
The Steriking® See-through Self-sealable pouches are intended for use as packing material for medical devices in sterilization by steam, ethylene oxide gas, or by formaldehyde in health care establishments. The common steam sterilization conditions are 3 min at 134 °C or 15 min at 121 °C.
  • For single-use only
The Steriking® See-Through packages are constructed of medical grade paper (70 g/m²) that is heat-sealed together with a multiply PET/PP-plastic laminate (12/40 µm). Pouches are intended for closing tightly with adhesive strip according to instructions printed.
The paper is a high-weight medical grade with improved barrier and water repellent properties. The controlled pore size provides for effective air evacuation and steam penetration. The specially treated surface facilitates strong sealing against the film but allows fiber-free peeling off without breaks. The paper conforms to the requirements of
the European EN 868-3:2009 and it is free from dirt, toxic substances and odor.
Certifications: Conform to the international product standards and norms: ISO 11607-1:2006, ISO 11607-2:2006 and EN 868-5:2009.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty