Would you like to log in again?
Dispensing Needles, OK International
Supplier: OK International
TE714100PK TE725050PK TE723050PK TE730025PK TE718150PK TE714150PK TE715050PK TE714150B45PK TE727050PK TE722050B90PK TE718050PK TE723050B45PK TE722050PK TE720150PK TE718050B45PK TE725025PK TE721150B45PK TE732025PK TE722150PK TE721050PK TE714050PK TE720100PK TE725050B45PK TE720025PK TE722100PK TE723100PK TE727050B45PK TE730050PK TE714050B45PK TE720050PK TE722050B45PK TE720050B45PK
76239-358PK 83.96 USD InStock
76239-358 76239-854 76239-356 76239-378 76239-852 76239-858 76239-856 76239-384 76239-362 76239-360 76239-382 76239-860 76239-366 76239-388 76239-386 76239-364 76239-380 76268-288 76239-864 76239-862 76239-368 76239-868 76239-866 76239-372 76239-392 76239-370 76239-376 76239-354 76239-398 76239-396 76239-374 76239-390
Dispensing Needles, OK International
Syringe Needles
The TE Series Premier dispensing tips consist of a stainless steel cannula with a double Helix polypropylene hub, for secure syringe barrel attachment. TE needles with Precision bent at 45̊ and 90̊ angles are perfect for dispensing fluid in hard to reach places.
  • Maximum operating pressure at 100 psi
This premier line of dispensing tips is burr-free, and has an electro-polished cannula for unobstructed and consistent material flow. In addition they are silicone and chloride-free.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty