Would you like to log in again?
Dispensing Tips, OK International
Supplier: OK International
TT20-DHUV-PK TT18-DHUV-PK TT22-DHUV-PK TT25-DHUV-PK TT16-DHUV-PK TT14-DHUV-PK TE-TT27-DHUV
76239-420PK 83.55 USD InStock
76239-420 76239-424 76239-556 76239-422 76239-418 76240-408 76239-416
Dispensing Tips, OK International
Syringe Needles
The tapered tip series designed with a double helix threads and universal luer lock to provide secure connection to industrial syringes or dispense valves.
  • Silicone and chloride-free with a maximum operating pressure 100 psi (6.9 bar)
  • Flash free orifice ensures accurate and repeatable dispensing
These tips are recommended for applications where dispensing medium and high-viscosity fluids with a high degree of precision and consistency is required. A smooth internal fluid path designed reduces turbulence flow, prevents air entrapment and fluid dripping caused by back pressure built up.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty