Would you like to log in again?
Metal Expansion Wristband, SCS, Desco Industries
Supplier: Desco Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2205
76239-632EA 55.65 USD InStock
76239-632
Metal Expansion Wristband, SCS, Desco Industries
Grounders Wrist Grounders
Metal Expansion Wristband made from stainless steel, the durable premium performance metal wristband creates a reliable electrical contact to the user’s wrist.
  • Durable and comfortable
Each link is covered with an insulative plastic cap. Use with single-wire coil cord with 4 mm female socket snap.
Certifications: Meets ANSI/ESD S20.20 tested per ANSI/ESD S1.1 and ESD TR53.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty