Would you like to log in again?
DisCide Ultra Towelettes, Palmero Healthcare
Supplier: PALMERO HEALTHCARE TE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
60DIS
76239-638CS 347.55 USD InStock
76239-638
DisCide Ultra Towelettes, Palmero Healthcare
Wipes Disposable Wipes and Towels Disinfectant Wipes
This is a hospital-level, one-step, ready-to-use quaternary ammonium, intermediate high-level alcohol-based disinfectant.
  • Container/lid does not contain latex
  • Non-corrosive and non-staining with pleasant herbal scent
  • No unsightly residue
This is proven to kill deadly pathogens in one minute or less for accelerated operatory turnover.
Certifications: FDA and EPA registered.
Ordering information: Available in towelettes, spray, or bulk gallon refill.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty