Would you like to log in again?
Power Rails, Production Basics
Supplier: Production Basics
PB-8321 PB-8320 PB-8323 PB-8324 PB-8326 PB-8327 PB-8322 PB-8319
76239-064EA 394.42 USD InStock
76239-064 76239-062 76240-288 76239-058 76239-660 76240-290 76239-060 76240-292
Power Rails, Production Basics
Furniture Casework
The Power Rail provides integrated power outlets for the workstation. The slim profile provides more room for accessories and has a sleek look to match the modern facility.
  • Mounts to frame or uprights
  • A lighted control switch includes a 20 A or 15 A circuit breaker
  • 36" and 48" Power Rails feature 6 outlets
  • 60" and 72" Power Rails feature 8 outlets
  • Black wrinkle epoxy powder coat finish matches the other workbench accessories
Delivery information: Supplied with two sets of brackets that allow to mount the power rail to frame/uprights or to the worksurface of the bench.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty