Would you like to log in again?
Xcelite® General Purpose Shear Cutter, Apex
Supplier: Apex
Xcelite®
XC-170MN 1178MN
76239-690EA 55.46 USD InStock
76239-690 76239-688
Xcelite® General Purpose Shear Cutter, Apex
Cutters
These low-profile general-purpose shear cutters are providing superior by-pass shear cutting action.
  • Better cuts with half the effort
  • Flush cuts soft wire up to 20 AWG
Greatly reduced mechanical shock delivered to the work.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty