Would you like to log in again?
Clamp Gaskets, Garlock Hygienic Technologies
Supplier: Garlock
40MPSFY-250 42MPSFY-050
76239-916BX 7.48 USD InStock
76239-916 76239-914
Clamp Gaskets, Garlock Hygienic Technologies
Seals Gaskets
These gaskets are used in processing pipeline systems for the food, dairy, beverage, pharmaceutical and bio tech industries.
Certifications: Meets criteria of the FDA, and accepted by the USDA, and USP Class VI certifications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty