Would you like to log in again?
AciTek® 49-252, Cleanroom Acid, Gloves, Ansell
Supplier: Ansell
AciTek®
174148
76239-936CS 456.48 USD InStock
76239-936
AciTek® 49-252, Cleanroom Acid, Gloves, Ansell
Gloves
These hand-specific, powder-free gloves are made from 100% natural latex polymer, which offers good protection from a wide spectrum of acids, etchants, and solvents.
  • Hand-specific
  • Powder-free
  • Straight cuffs
The ergonomic design, featuring a straight cuff, provides exceptional flexibility and comfort for extended wearing. Gloves are chlorinated inside and outside. Fishscale texture on fingers and palms provides a sure hold on glassware and other smooth or slippery surfaces.
Certifications: Meets EN 374 standards.
Packaging: Gloves are packaged in an ISO Class 5 (FED-STD-209E Class 100/M3.5) environment. Cleanroom-compatible packaging features easy tear-open configuration and contains one pair per inner poly bag and 12 pairs per outer poly bag. Lot number and size label are printed on inner poly bag in IPA-resistant ink.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty