Would you like to log in again?
Three Layer Vinyl Mat Roll, SCS Division of Desco
Supplier: Desco Industries
8234 8264 8274
76240-010EA 926.1 USD InStock
76240-010 76245-482 76248-260
Three Layer Vinyl Mat Roll, SCS Division of Desco
Floor Mats Tacky Mats
These three layer vinyl mat of two types with or without grounding hardware, provides reliable path to ground and eliminates static charge generation as a worksurface table mat, shelf liner and cart liner.
Provides main discharge path to ground and durable, nonskid cushion, choice for components susceptible to physical shock.
Certifications: Meets ANSI/ESD S20.20 ANSI/ESD S4.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty