Would you like to log in again?
Swabs, FG Clean Wipes
Supplier: FG Cleanwipes
6S144 6S160 6S160
76240-388CS 4546.99 USD InStock
76240-388 76238-284 76238-285
Swabs, FG Clean Wipes
Swabs Swabs for Cleaning
Polyurethane foam tip swabs with Polypropylene handle.
  • Polyurethane foam head provides good particle entrapment and removal
  • Ideal for cleaning with solvents with the exception of ketones
The super-absorbent foam heads offer the perfect swab for nearly any industry or application.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty