Would you like to log in again?
Lazer Line™ 7000 Angulated Head Cutters, Excelta
Supplier: Excelta Corporation
7204E 7182E 7282E
76246-296EA 312.9 USD InStock
76246-296 76240-402 76240-072
Lazer Line™ 7000 Angulated Head Cutters, Excelta
Cutters
These ultra-precision 7000 series fine tip cutters are made from 1% carbon tool steel.
  • 7100 series: Long cushion grips, ESD safe
  • 7200 series: Molded grips, ESD safe
The cutting edges are induction hardened to 63-65 Rockwell C. The redesigned, longer lasting, stainless steel leaf springs provide smooth operation. The CNC machining allows the joint surfaces to be perfect to allow for long life.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty