Would you like to log in again?
Composite Wipes, FG Clean Wipes
Supplier: FG Cleanwipes
7740844L00
76240-534CS 287.78 USD InStock
76240-534
Composite Wipes, FG Clean Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Multilayered, outer polypropylene layers thermally bonded to cellulose core, with innovative cleaning properties.
  • Outer layers resist abrasion and wick rapidly
  • Inner layer traps and holds liquids
  • Delivers dryer wiper surface wiping alongside super absorbency
  • Strong, durable and absorbent
Ideal for cleaning acids and other harmful liquids. Low particulate and chemical extractables.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty