Would you like to log in again?
HyFlex® 70-215 Cut resistant gloves, Ansell
Supplier: Ansell
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyFlex®
222122 222124
76240-570CS 1787.17 USD InStock
76240-570 76240-572
HyFlex® 70-215 Cut resistant gloves, Ansell
Gloves
The 100% dupont kevlar® construction gloves offer very high cut resistance to weight ratio, use of Terry loop technique enhances heat insulation.
  • Protection from sharp edges of metals, ceramics, glass and other materials
  • Reversible design allows glove to be worn on either hand
  • Launderable
The reinforced thumb welt increases abrasion resistance also adding to glove durability. The ambidextrous gloves are fully washable and can be re-used many times.
Packaging: Also available in vend pack.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty