Would you like to log in again?
Horse Hair Acid Brush, Torrington Brush Works
Supplier: Torrington
13011
76240-592EA 1 USD InStock
76240-592
Horse Hair Acid Brush, Torrington Brush Works
Brushes
Standard horse hair acid brush with the handle of bright tin plated.
  • Ferrule is double crimped and has a staple set to hold the filament securely in place
Used for applying glue, soldering paste and other types of acidic applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty