Would you like to log in again?
ESD Paper, ALX Technical
Supplier: ALX TECHNICAL TE
A09-1711-2BT38
76240-598CS 395.31 USD InStock
76240-598
ESD Paper, ALX Technical
Paper
This static dissipative (ESD) paper eliminates static quickly.
  • Does not rely on humidity
  • 8.5×11"
Consistantly within the static dissipative range of 107 to 109.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty