Would you like to log in again?
Essentra Presaturated Wipes, FG Clean Wipes
Supplier: FG Cleanwipes
6LS964685
76240-762CS 854.57 USD InStock
76240-762
Essentra Presaturated Wipes, FG Clean Wipes
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
This wipes to control contamination in critical environments that happened on wet cleaning.
  • Reduced VOCs
  • Minimized bulk liquid storage
  • Safer to ship, store and use
  • Consistency in application of solvent
Purity and cleanliness to support ISO 4 and ISO 5 environments.
Packaging: Canisters with spring-loaded lids.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty