Would you like to log in again?
Semco® Legacy Red Scraper, Bergdahl Associates
Supplier: BERGDAHL ASSOCIATES INC. TE
234350
76240-836CS 1596.71 USD InStock
76240-836
Semco® Legacy Red Scraper, Bergdahl Associates
Scrapers
This scraper is fabricated from Celcon® POM and will not scratch or score aluminum substrates.
Certifications: The tool has been evaluated and approved by Boeing for the removal of cured sealant and is listed in Boeing BAC 5000 process specification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty