Would you like to log in again?
AseptiQuik® C Connectors, Colder Products Company
Supplier: CPC (Colder Products Company)
CPC® AseptiQuik® C
AQC33012HT AQC44012HT AQC22008
76253-976EA 52.25 USD InStock
76253-976 76240-850 76240-848
AseptiQuik® C Connectors, Colder Products Company
Fittings and Connectors
AseptiQuik® C Connectors provide quick and easy sterile connections, even in non-sterile environments. The 'Click-Pull-Twist' design enables users to transfer media easily with less risk of operator error.
  • Operating temperature: 4 to 40 °C
  • Ensure simultaneous and secure removal of both membranes
  • Audible confirmation of completed assembly steps
  • Open access through multiple supply chain partners
  • Secures final connection preventing disassembly
Biopharmaceutical manufacturers can make sterile connections with the quality and market availability they expect from the leader in single-use connection technology.
Certifications: USP class VI, ADCF certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty