Would you like to log in again?
PLC Series Connector, Colder Products Company
Supplier: CPC (Colder Products Company)
PLC120M10
76240-878PK 472.68 USD InStock
76240-878
PLC Series Connector, Colder Products Company
Fittings and Connectors
The 1/4" flow PLC Series is proven worldwide in thousands of applications and offers the widest selection of sizes and configurations. PLC quick disconnect coupling is injection molded from acetal thermoplastic and resistant to most mild chemical solutions.
  • Operating temperature: −40 to 82 °C continuous
  • Assurance of a reliable connection
  • Fewer leak points, shorter assemblies and faster installations
This coupling provides the flexibility of multiple configurations and terminations and mate with both LC metal and PLC12 polypropylene couplings.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty