Would you like to log in again?
Bevco® Everlast ESD Polyurethane Chairs
Supplier: BevCo Precision Manufacturing
Bevco®
7352E-CARD5S-AAE 7050E-CARD5S 7551E
76241-334EA 3754 USD InStock
76241-334 76240-942 76249-754
Bevco® Everlast ESD Polyurethane Chairs
Chairs
Protect Your Equipment with an ESD Chair.
  • The black ESD polyurethane seat and back are static dissipative and are impervious to water, oil, and most chemicals
  • Polyurethane is puncture-resistant and easy-to-clean
  • Contoured back is fully adjustable encouraging proper spinal alignment
  • Convenient handle makes moving the chair effortless
  • Waterfall seat aids in lower body circulation, improving overall wellness
  • Pneumatic height adjustment allows you to easily change your seat height
  • Sturdy polished aluminum base comes witha brass drag chain for an additional grounding point
  • 300 lb. weight capacity
  • ESD environment
ESD chairs are designed to minimize static discharge that is generated while sitting and to safely discharge it through the floor grounds. This helps protect your delicate equipment from damage. Bevco's Everlast electrostatic dissipative (ESD) or static control chairs meet ANSI/ESD STM12.1–2019 standards providing fully grounded peace of mind. By making a Bevco ESD chair the first line of defense in your ANSI/ESD S20.20-2021 program, you can ensure maximum protection for your equipment.
Certifications: ANSI/BIFMA; GreenGuard Gold.
Ordering information: 12-year warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty