Would you like to log in again?
Mopheads, Carlisle Sanitary Maintenance
Supplier: CARLISLE SANITARY MAINT. PROD TE
369552B00
76240-974CS 333.25 USD InStock
76240-974
Mopheads, Carlisle Sanitary Maintenance
Mops
Looped-end construction provides greater floor coverage versus a cottoncut-end mop.
  • 5" color-coded headband
  • 4-ply looped-end cotton mops are good for aggressive cleaning and absorbancy
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty