Would you like to log in again?
Ergo-Zoom® Microscope with Standard Ball Bearing Base, O.C. White
Supplier: O.C. White
TKEZ-850-L-LV2
76241-178EA 15415.52 USD InStock
76241-178
Ergo-Zoom® Microscope with Standard Ball Bearing Base, O.C. White
Microscopes
This microscope with scientific grade common main objective (CMO) and optical design delivers distortion free, high resolution flat field images.
  • Modular microscope outfitted to match exact needs
  • Features detented zoom stops for repeatable magnification location
  • Lens (1×) with working distance and focal length of 78 mm, field of view 30 to 4.8 mm
For inspecting mission-critical parts, there is no finer solution.
The precision glide ball bearing arm gives effortless motion with 20" of horizontal movement and rock solid stability, built with ESD safety.
Swiveling light source: 24 V lamp, 100 to 240 V supply.
Delivery information: Includes double ball bearing base, wide 10× eyepiece, 1× auxillary lens, ESD safe, LED light ring.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty