Would you like to log in again?
Cleanroom Wipes, FG Clean Wipes
Supplier: FG Cleanwipes
97177WEAG00 78520 97177BEAG00 97177YW8A00 97177GN8A00 97177YW8A00 97177WE8A00 97177PK8A00
76244-882CS 829.09 USD InStock
76244-882 76238-322 76244-876 76244-888 76241-202 76239-984 76238-324 76238-326
Cleanroom Wipes, FG Clean Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
The paper and notebooks are specially designed for use in cleanrooms. Each page is impregnated and coated with a unique polymer formulation that inhibits particulate generation and chemical extractables. Control contamination, make cleaning easier, and increase product yield with cleanroom compatible paper.
  • Ultralow particle generation
  • Ultralow metallic ion content
  • High brightness and opacity
  • High tear and tensile strength
  • Heat resistant
  • Compatible with virtually any ink system
The wipes keeps critical environments clean reducing the risk of contamination before it can even be a factor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty