Would you like to log in again?
3M™ Electrically Conductive Connector Covers, 4270 Series, Menda
Supplier: Menda
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
34219 34210 34218 34216 34209 34221 34217
76241-318CS 4419.15 USD InStock
76241-318 76241-316 76241-314 76241-322 76241-312 76241-320 76241-484
3M™ Electrically Conductive Connector Covers, 4270 Series, Menda
Fittings and Connectors
Permanently volume conductive and free of carbon black particulate. The connector covers do not leave black marks and are more suited to clean situations.
  • Noncorrosive
  • Conformability
The polyethylene material used in the covers was selected for its resilience. Properly sized, the covers provide a snug, secure fit. This helps keep connectors clean and prevents the cover from accidentally falling off.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty