Would you like to log in again?
Micro Tweezers, JBC Tools USA
Supplier: JBC
PA120-A
76242-012EA 398.41 USD InStock
76242-012
Micro Tweezers, JBC Tools USA
Forceps
This microtweezer is designed for soldering and desoldering SMD components easily, reducing handling time.
  • Ergonomics and the short distance tip-to-grip also ensure greater accuracy and comfort when working
  • Enables to align the cartridges in order to adapt them to the component
This tweezer works with two cartridges managed individually by the control unit to ensure the maximum accuracy and a quick temperature recovery. Each cartridge can deliver up to 40 W.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty