Would you like to log in again?
Statshield® Metal-In Bag, Menda, Desco Industries
Supplier: Desco Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
13500 13465
76242-026BX 106.3 USD InStock
76242-026 76245-464
Statshield® Metal-In Bag, Menda, Desco Industries
Bags Storage Bags
Transparent metal in standard ESD shielding bags.
  • 40% light transmission
  • Integral antistatic, low tribo-charging properties
  • Polycarbonate compatible containing no amines or N-octanic acid
Printed with ESD Protective Symbol and Lot Code Information. Allows bag to be used throughout an ESD Protected Area and grounded when placed on a grounded surface or handled by a grounded operator.
Certifications: Meets ANSI/ESD S20.20 and ANSI/ESD S541 requirements for ESD shielding packaging inside and outside an ESD Protected Area (EPA). Lead-free RoHS compliant.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty