Would you like to log in again?
Edsyn™ Soldering Tip Cleaner, Jensen Tools
Supplier: Jenson Tools
EA439-1
76242-486EA 22.33 USD InStock
76242-486
Edsyn™ Soldering Tip Cleaner, Jensen Tools
Soldering Supplies Desoldering Tools
Soft wire mesh design cleans tips without using water and will not reduce tip temperature as with wet sponges.
  • Never needs water
Instead of using a wiping motion as with regular sponges, just thrust the iron tip straight into the mesh and remove.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty