Would you like to log in again?
Removable Repositionable Tape 666, 3M
Supplier: 3M
666-2X72YD
76242-714EA 238.67 USD InStock
76242-714
Removable Repositionable Tape 666, 3M
Tapes
Removable Repositionable Tape 666 is a low density polyethylene (LDPE) liner with repositionable adhesive 1070 on a clear UPVC film carrier.
  • Slight side-to-side adhesion differential
  • Good adhesive anchorage to the film carrier
Slight differential tack. Low temperature and medium solvent resistance. Medium HSE and medium LSE. Medium tack allows for removal without residue from many films, foils, papers and is ideal for applications where repositioning is desired.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty