Would you like to log in again?
Adhesive Transfer Tape 9485PC, Tapecase
Supplier: TAPECASE LTD. TE
9485PC-1/4
76242-726EA 58.95 USD InStock
76242-726
Adhesive Transfer Tape 9485PC, Tapecase
Tapes
Adhesive Transfer Tape 9485PC is an excellent solution bonding high and low surface energy materials.
  • Adhesive is fiber reinforced for roll stability
  • Low odor for improved working environment
  • Excellent chemical resistance and shear and peel strength
Designed for short term temperature exposure up to 450 °F (232 °C) and long term 300 °F (149 °C). It can also be used for joining materials that have rougher surfaces.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty