Would you like to log in again?
Medical / Laboratory ESD-Safe Fabric Chair, Gibo Kodama
Supplier: GIBO KODAMA TE
EIN51IR-F851-07
76243-104EA 3803.2 USD InStock
76243-104
Medical / Laboratory ESD-Safe Fabric Chair, Gibo Kodama
Chairs
Adjustable ESD safe fabric chair with aluminum base and saddle seat.
  • Autonomous tilt control with seat and back angle adjustments
  • Chrome plated cylinder column
  • Saddle seat
Surface resistance: 106 Ω/sq
Seat adjustable from 17 - 211/2"
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty