Would you like to log in again?
Xcelite® Pro-Series Electronic Screwdrivers, Apex
Supplier: Apex
Xcelite®
XPE102 XPE5324 XPE144 XPE101 XPE3166 XPE184
76243-412EA 18.42 USD InStock
76243-412 76243-124 76244-380 76243-122 76243-112 76243-120
Xcelite® Pro-Series Electronic Screwdrivers, Apex
Screwdrivers
These screwdrivers feature hard shockproof plastic handle core for durability, while the outer coating is soft rubber for comfort and slip resistance.
  • ESD safe hard plastic core to protect sensitive components
  • Narrow pinch grip for comfort and control
  • Thumb bolster allows added downward force
  • Satin finish chrome molybdenum steel shafts provide durability
  • Specially treated black oxide tips eliminate light reflection
The handles are ergonomically designed, allowing sustained high torque with minimal slippage and muscle fatigue. Designed with smooth shatterproof seamless handle cap for minimal friction between hand and handle when screwdriver is being turned.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty