Would you like to log in again?
ESD Tray Inlays, Intermetro Industries
Supplier: Intermetro
CBNTC-INLAY02
76243-228EA 87.41 USD InStock
76243-228
ESD Tray Inlays, Intermetro Industries
Trays/Dishes/Pots Laboratory Trays/Dishes
Provides soft-surface inlay for cushioned PCB transport. Inlay provides static dissipative ESD protection.
Not recommended for use in soldering and assembly applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty