Would you like to log in again?
Steam-Thru II Configuration Coupling, Colder Products Company
Supplier: CPC (Colder Products Company)
STC2021000
76243-334PK 1929.47 USD InStock
76243-334
Steam-Thru II Configuration Coupling, Colder Products Company
Fittings and Connectors
Steam-Thru II Connections offer the flexibility of "steam on" and "steam off" functionality. The innovative design allows the valve to be returned to the steam position enabling a second SIP cycle following media transfer.
Materials of construction include USP class 6 animal free (BSE/TSE) polysulfone, USP class 6 andimal free (BSE/TSE) silicone orings and seals.
Certifications: USP Class VI, BSE/TSE free materials (ADCF)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty