Would you like to log in again?
3M™ F9469PC VHB™ Adhesive Transfer Tape, Tapecase
Supplier: TAPECASE LTD. TE
F9469PC-12
76243-410EA 1412.4 USD InStock
76243-410
3M™ F9469PC VHB™ Adhesive Transfer Tape, Tapecase
Tapes
3M™ Adhesive Transfer Tapes are rolls of pressure sensitive adhesive pre-applied to a special release liner. For application, the tape is simply pressed, adhesive side down, to a surface and the liner is peeled off.
  • High holding power in static and dynamic loads
  • Suitable for industrial joining and metal fabrication
  • Eliminates mechanical fastening in many jobs
  • High performance acrylic adhesive
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty