Would you like to log in again?
Polyester Film Tape 850, 3M
Supplier: 3M
7100077066 7000123285 7100065924 7000035828 7100065924
76268-322CS 3409.63 USD InStock
76268-322 76243-644 76242-720 76243-326 76242-718
Polyester Film Tape 850, 3M
Tapes
Polyester Film Tape 850 is generally used for splicing and web applications. The backing is a thin, high-strength polyester which is abrasion resistant to help reduce splice failures while resisting breakdowns during rough handling and web processing.
  • Low modulus, low Shore A Hardness
  • Thin, high strength polyester film
  • Aggressive acrylic adhesive
Coated with an aggressive pressure sensitive acrylic adhesive, this tape grabs on contact and holds securely. It meets low-caliper requirements to ensure it won't interfere with most machinery. The tape can withstand a wide temperature range from –60 °F/–50 °C to 300 °F/150 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty