Would you like to log in again?
Proto® Reversible Safety Wire Twister Plier, Straight Jaw, ORS Nasco
Supplier: ORS Nasco
577-196
76243-676EA 124.97 USD InStock
76243-676
Proto® Reversible Safety Wire Twister Plier, Straight Jaw, ORS Nasco
Pliers
This plier is suitable for cutting stainless steel wire.
  • Heat-treated cutting edges
  • Automatic spring return speeds operation and reduces fatigue
  • Right and left twist capability
  • Holds wire securely and resists kinking
  • Easy access to confined, restricted areas
Aviation and other applications where vibration may cause fasteners to back out.
Certifications: Meets SAE Aerospace Standard AS567.
Caution: Cancer and reproductive harm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty