Would you like to log in again?
Replaceable Fiber Tipped Tweezers, Lindstrom Precision Tools
Supplier: Williams
259CFR-SA 259CF-SA 259-ACF
76240-430EA 35.35 USD InStock
76240-430 76243-682 76240-096
Replaceable Fiber Tipped Tweezers, Lindstrom Precision Tools
Forceps
Polished stainless steel body with replaceable plastic tips. ESD carbon fiber tips do not leave metal marks and grant a contamination free performance.
  • Easy tip replacement system
  • Replacement tips include two tips and two screws in a plastic bag
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty