Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Mantis Elite stereo microscope, Vision Engineering
Supplier: Vision Engineering
MES-005 MEH-001
76243-720EA 8484.5 USD InStock
76243-720 76243-836
Mantis Elite stereo microscope, Vision Engineering
Microscopes
Mantis Elite is an ergonomic eyepieceless stereo microscope, offering superb optical performance with magnification options up to 20x, making it a perfect alternative to more traditional inspection stereo microscopes.
  • 2x – 20x magnification options (2 in quick change turret)
  • Superior ergonomics for fatigue-free viewing and increased quality / productivity
  • Superb hand-eye co-ordination for inspection and manipulation tasks
  • Long working distance and large field of view
  • True colour, LED illumination providing up to 10,000 hours of shadow-free viewing
Mantis Elite is a patented ‘eyepieceless’ stereo microscope, offering truly superb 3-D imaging and unparalleled levels of comfort, reducing operator fatigue and increasing quality and productivity. Winner of numerous industry design awards, the outstanding ergonomic design is unrivalled by other inspection stereo viewers.
Patented optical technology removes the need for restrictive eyepieces of a conventional microscope, allowing operators freedom of head movement, providing superb ergonomics and hand-eye co-ordination, plus the ability to wear glasses, if required.
Mantis Elite is a perfect alternative to traditional stereo microscopes, in use throughout the world for a wide range of inspection, preparation and manipulation tasks requiring hand-to-eye co-ordination.
Ordering information: Head and stand need to be ordered separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty