Would you like to log in again?
Anti-Static Bags, Elkay Plastics
Supplier: Elkay Plastics
SSR1624
76244-368CS 651.48 USD InStock
76244-368
Anti-Static Bags, Elkay Plastics
Bags Storage Bags
Bags with buried metal construction provides double protection against static discharge. Amine/amide-free interior will not corrode polycarbonate surfaces and produces no outgassing.
  • The semi-transparent material allows for easy identification of contents
  • Great for protecting contents that must be accessed multiple times
The ⅜" side seal ensures packaging integrity during storage and handling. Defect prevention versus defect detection builds quality into every bag.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty