Would you like to log in again?
Static Shielding Bags, 1000 Series, Z-Mar Technology
Supplier: Z-Mar Technology
TRILAY8.5X11QP TRILAY8X8QPLZ TRILAY8X8QPLZ TRILAY6X6QPLZ TRILAY12X16QPL
76244-783PK 1365.62 USD InStock
76244-783 76244-784 76244-776 76244-780 76244-782
Static Shielding Bags, 1000 Series, Z-Mar Technology
Bags Storage Bags
These transparent metal-in static shielding bags are designed to provide a static safe environment for sensitive electronic devices.
  • Heat sealable bags
  • Bags are printed with an ESD protective symbol and a lot code for traceability
These bags are manufactured from a polyester, metal, polyethylene laminate. The polyester dielectric in concert with the metal layer provides discharge shielding. The exterior being static dissipative allows electrostatic charges to be removed when grounded.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty