Would you like to log in again?
Tape 8402, Polyester, 3M
Supplier: 3M
7000048552
76244-964CS 5320.48 USD InStock
76244-964
Tape 8402, Polyester, 3M
Tapes
Polyester Tape 8402 is a thin, conformable polyester tape ideal for masking in high temperature applications and adheres well to silicone coated surfaces.
  • Ideal for splicing applications on surfaces
  • Green color makes for easier identification
  • Chemical resistant adhesive
he tape has a silicone adhesive that functions well for masking and removes cleanly, in one piece, from most surfaces. This tape also excels in permanent splicing operations on silicone liners, papers and films subject to high temperatures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty