Transparent, metal-in static shielding bags are designed to provide a static safe environment for ESD sensitive electronic devices.
- Protects ESD sensitive contents from electrostatic fields and electrostatic discharges (ESD)
- Metal-In film laminate is 0.0028" thick (2.8 mil)
- Dissipative Inner and Outer Surfaces Rs 1×10⁴ to <1×10¹¹ ohms per ANSI/ESD STM11.11 and Static Control Bag ANSI/ESD S11.4 Level 3
- Bags can be used throughout an ESD Protected Area and grounded when placed on a grounded surface or handled by a grounded operator
Bags are printed with an ESD protective symbol and a lot code for traceability. Open top and ZipTop styles are available. The bags are heat sealable. SCS Static Shielding Bags are manufactured from a polyester, metal, polyethylene laminate. The polyester dielectric, in concert with the metal layer, provides discharge shielding. The exterior being static dissipative allows electrostatic charges to be removed when grounded.
Certifications: Meets ANSI/ESD S20.20 and ANSI/ESD S541 requirements for ESD shielding packaging inside and outside. Lead-free RoHS 2, REACH, and Conflict Minerals Compliant.
<10 nJ Discharge Shielding Energy Limit Test per ANSI/ESD STM11.31.